missing translation for 'onlineSavingsMsg'
Learn More

Thieno[3,2-b]thiophene 98.0+%, TCI America™

Catalog No. T25135G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25135G 5 g
T25131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25135G Supplier TCI America Supplier No. T25135G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 251-41-2
Molecular Formula C6H4S2
Molecular Weight (g/mol) 140.218
MDL Number MFCD00179479
InChI Key VJYJJHQEVLEOFL-UHFFFAOYSA-N
Synonym thieno 3,2-b thiophene, 1,4-dithiapentalene, thienothiophene, 1,4-thiophthene, pubchem21292, acmc-209ghi, thieno 3,2-b thiophen, thieno 3,2-b-thiophene
PubChem CID 136063
IUPAC Name thieno[3,2-b]thiophene
SMILES C1=CSC2=C1SC=C2

Specifications

Melting Point 56°C
Boiling Point 71°C
Quantity 5 g
Formula Weight 140.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Thieno[3,2-b]thiophene