accessibility menu, dialog, popup

UserName

H3COONOO

CAS RN 2483-57-0

IUPAC Name: methyl 2-nitroacetate
Synonyms: nitroacetic acid methyl ester
Molecular Weight (g/mol): 119.08
Molecular Formula: C3H5NO4
InChi Key: ALBSWLMUHHZLLR-UHFFFAOYSA-N
SMILES: COC(=O)C[N+]([O-])=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)