missing translation for 'onlineSavingsMsg'
Learn More

Methyl nitroacetate, 97+%

Catalog No. AAL1895009
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1895009 10 g
AAL1895004 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1895009 Supplier Thermo Scientific Chemicals Supplier No. L1895009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2483-57-0 | C3H5NO4 | 119.076 g/mol

Methyl nitroacetate is used in the synthesis of aromatic and heteroaromatic linear (E)-alfa-nitro-arylpentenoates. It is also used to prepare phenyliodonium ylide. Further, it is utilized in the highly enantioselective and diastereoselective copper(I)-catalyzed cyclopropanation of alkenes. In addition to this, it is employed in the production of methyl (Z)-2-nitro-3-(4-nitrophenyl)-2-propenoate through reaction with 4-nitrobenzylideneaniline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl nitroacetate is used in the synthesis of aromatic and heteroaromatic linear (E)-alfa-nitro-arylpentenoates. It is also used to prepare phenyliodonium ylide. Further, it is utilized in the highly enantioselective and diastereoselective copper(I)-catalyzed cyclopropanation of alkenes. In addition to this, it is employed in the production of methyl (Z)-2-nitro-3-(4-nitrophenyl)-2-propenoate through reaction with 4-nitrobenzylideneaniline.

Solubility
Miscible with ethanol, ether and most organic solvents.

Notes
Light sensitive. Incompatible withstrong bases.

Chemical Identifiers

CAS 2483-57-0
Molecular Formula C3H5NO4
Molecular Weight (g/mol) 119.076
MDL Number MFCD00075480
InChI Key ALBSWLMUHHZLLR-UHFFFAOYSA-N
Synonym methyl nitroacetate, nitroacetic acid methyl ester, acetic acid, nitro-, methyl ester, methylnitroacetate, methyl nitrozcetate, methyl nitro acetate, zlchem 149, methyl a-nitroacetate, methyl 2-nitroethanoate, acmc-1clk8
PubChem CID 17206
IUPAC Name methyl 2-nitroacetate
SMILES COC(=O)C[N+](=O)[O-]

Specifications

Density 1.294
Boiling Point 195°C to 198°C
Flash Point 105°C (221°F)
Odor Ester-like
Refractive Index 1.426
Linear Formula O2NCH2CO2CH3
Quantity 10 g
Beilstein 1758670
Sensitivity Light sensitive
Solubility Information Miscible with ethanol,ether and most organic solvents.
Formula Weight 119.08
Percent Purity 98%
Chemical Name or Material Methyl nitroacetate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 219-622-0
RTECSNumber AJ1093000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only