missing translation for 'onlineSavingsMsg'
Learn More

Methyl Nitroacetate 97.0+%, TCI America™

Catalog No. N06565G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06565G 5 g
N065625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06565G Supplier TCI America Supplier No. N06565G

Chemical Identifiers

CAS 2483-57-0
Molecular Formula C3H5NO4
Molecular Weight (g/mol) 119.076
MDL Number MFCD00075480
InChI Key ALBSWLMUHHZLLR-UHFFFAOYSA-N
Synonym methyl nitroacetate, nitroacetic acid methyl ester, acetic acid, nitro-, methyl ester, methylnitroacetate, methyl nitrozcetate, methyl nitro acetate, zlchem 149, methyl a-nitroacetate, methyl 2-nitroethanoate, acmc-1clk8
PubChem CID 17206
IUPAC Name methyl 2-nitroacetate
SMILES COC(=O)C[N+](=O)[O-]

Specifications

Color Yellow
Boiling Point 100°C
Quantity 5 g
Formula Weight 119.08
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Nitroacetate
RTECSNumber AJ1093000
TSCA No