accessibility menu, dialog, popup

UserName

CAS RN 24517-64-4

NH2NHN

CAS RN 24517-64-4

IUPAC Name: 2-amino-3-cyanopyridin-1-ium
Synonyms: 2-amino-3-cyanopyridin-1-ium
Molecular Weight (g/mol): 120.13
Molecular Formula: C6H6N3
InChi Key: YYXDQRRDNPRJFL-UHFFFAOYSA-O
SMILES: NC1=[NH+]C=CC=C1C#N
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)