missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-cyanopyridine 98.0+%, TCI America™

Catalog No. A24435G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24435G 5 g
A244325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24435G Supplier TCI America Supplier No. A24435G

Chemical Identifiers

CAS 24517-64-4
Molecular Formula C6H6N3
Molecular Weight (g/mol) 120.13
MDL Number MFCD00128867
InChI Key YYXDQRRDNPRJFL-UHFFFAOYSA-O
Synonym 2-amino-3-cyanopyridine, 2-aminonicotinonitrile, 2-amino-3-pyridinecarbonitrile, 2-amino-pyridine-3-carbonitrile, 2-amino-nicotinonitrile, 3-pyridinecarbonitrile, 2-amino, 3-cyanopyridin-2-amine, 3-cyanopyridine-2-amine, aminonicotinonitrile, zlchem 506
PubChem CID 818259
IUPAC Name 2-amino-3-cyanopyridin-1-ium
SMILES NC1=[NH+]C=CC=C1C#N

Specifications

Melting Point 135°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 119.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-cyanopyridine