missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-cyanopyridine, 98%

Catalog No. AAL1961806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1961806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1961806 Supplier Thermo Scientific Chemicals Supplier No. L1961806

CAS: 24517-64-4 | C6H6N3 | 120.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24517-64-4
Molecular Formula C6H6N3
Molecular Weight (g/mol) 120.13
MDL Number MFCD00128867
InChI Key YYXDQRRDNPRJFL-UHFFFAOYSA-O
Synonym 2-amino-3-cyanopyridine, 2-aminonicotinonitrile, 2-amino-3-pyridinecarbonitrile, 2-amino-pyridine-3-carbonitrile, 2-amino-nicotinonitrile, 3-pyridinecarbonitrile, 2-amino, 3-cyanopyridin-2-amine, 3-cyanopyridine-2-amine, aminonicotinonitrile, zlchem 506
PubChem CID 818259
IUPAC Name 2-amino-3-cyanopyridin-1-ium
SMILES NC1=[NH+]C=CC=C1C#N

Specifications

Melting Point 133°C to 135°C
Quantity 5 g
UN Number UN3439
Beilstein 115612
Formula Weight 119.13
Percent Purity 98%
Chemical Name or Material 2-Amino-3-cyanopyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only