accessibility menu, dialog, popup

UserName

CAS RN 2437-29-8

H3CNCH3CNCH3H3CH3CNCH3CNCH3H3CHOOOHOHOOOOHOOOO

CAS RN 2437-29-8

IUPAC Name: bis(bis[4-(dimethylamino)phenyl](phenyl)methylium) oxalic acid dihydrogen oxalate
Synonyms: bis(bis[4-(dimethylamino)phenyl](phenyl)methylium) oxalic acid dihydrogen oxalate
Molecular Weight (g/mol): 927.02
Molecular Formula: C52H54N4O12
InChi Key: CNYGFPPAGUCRIC-UHFFFAOYSA-L
SMILES: OC(=O)C(O)=O.OC(=O)C([O-])=O.OC(=O)C([O-])=O.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C
Quantity 
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
Color 
  • (7)
Grade 
  • (5)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (7)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (7)
  • (5)
  • (1)
  • (2)
  • (1)