missing translation for 'onlineSavingsMsg'
Learn More

Malachite Green, Oxalate 95.0+%, TCI America™

Catalog No. M049725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M049725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M049725G Supplier TCI America Supplier No. M049725G
Only null left

Chemical Identifiers

CAS 2437-29-8
Molecular Formula C52H54N4O12
Molecular Weight (g/mol) 927.02
MDL Number MFCD00011766,MFCD00151209
InChI Key CNYGFPPAGUCRIC-UHFFFAOYSA-L
Synonym malachite green oxalate, basic green 4, unii-gy1h9o33vu, gy1h9o33vu, malachite green, oxalate, dsstox_cid_5513, malachite green oxalate salt c.i. 42000, dsstox_rid_77817, dsstox_gsid_25513, n,n,n',n'-tetramethyl-4,4'-diaminotriphenylcarbenium oxalate
PubChem CID 2724411
IUPAC Name bis(bis[4-(dimethylamino)phenyl](phenyl)methylium) oxalic acid dihydrogen oxalate
SMILES OC(=O)C(O)=O.OC(=O)C([O-])=O.OC(=O)C([O-])=O.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C

Specifications

Color Green
Quantity 25 g
UN Number 2811
Formula Weight 927.02
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Malachite Green, Oxalate
EINECSNumber (5)-2033
RTECSNumber BQ1190000
TSCA No