missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Malachite Green oxalate

Catalog No. AAA1618622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1618622 100 g
AAA1618614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1618622 Supplier Thermo Scientific Chemicals Supplier No. A1618622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2437-29-8 | C52H54N4O12 | 927.02 g/mol

Biological stain, dye reagent, acid base indicator: pH 0.0 yellow, 2.0 green;11.6 green, 14 colorlessMalachite Green oxalate is used as the staining solution for embedded botanical material and as a spore stain. It is also used in studying the photocatalytic degradation with TiO2/WO3. It is employed for determining phosphate in serum and as a reference material in analytical chemistry for the quantification. It is also used as a pH indicator between pH 0.2 - 1.8. It is used as a dye for materials such as silk, leather, and paper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biological stain, dye reagent, acid base indicator: pH 0.0 yellow, 2.0 green;11.6 green, 14 colorlessMalachite Green oxalate is used as the staining solution for embedded botanical material and as a spore stain. It is also used in studying the photocatalytic degradation with TiO2/WO3. It is employed for determining phosphate in serum and as a reference material in analytical chemistry for the quantification. It is also used as a pH indicator between pH 0.2 - 1.8. It is used as a dye for materials such as silk, leather, and paper.

Solubility
Soluble in water, alcohol, methanol, ethanol and ethylene glycol monomethyl ether.

Notes
Incompatible with strong oxidizing agents, strong reducing agents, strong bases and strong acids.

Chemical Identifiers

CAS 2437-29-8
Molecular Formula C52H54N4O12
Molecular Weight (g/mol) 927.02
MDL Number MFCD00011766,MFCD00151209
InChI Key CNYGFPPAGUCRIC-UHFFFAOYSA-L
Synonym C.I. 42000
PubChem CID 2724411
IUPAC Name [4-[[4-(dimethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;2-hydroxy-2-oxoacetate;oxalic acid
SMILES OC(=O)C(O)=O.OC(=O)C([O-])=O.OC(=O)C([O-])=O.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C

Specifications

Melting Point 164°C (decomposition)
UN Number UN2811
Beilstein 3644550
Merck Index 14,5699
Solubility Information Soluble in water,alcohol,methanol,ethanol and ethylene glycol monomethyl ether.
Quantity 100 g
Formula Weight 927.02
Chemical Name or Material Malachite Green oxalate
Hazard Category H301-H318-H361d
Hazard Statement GHS H Statement
H318-H361-H302
Precautionary Statement P201-P202-P264b-P270-P281-P301+P310-P305+P351+P338-P308+P313-P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 219-441-7
RTECSNumber BQ1190000
TSCA Yes

RUO – Research Use Only