missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Malachite Green oxalate, pure, high purity biological stain

Catalog No. AC229785000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC229785000 500 g Glass bottle
AC229780250 25 g Glass bottle
AC229781000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC229785000 Supplier Thermo Scientific Chemicals Supplier No. 229785000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Malachite Green oxalate, C52H54N4O12, CAS Number-2437-29-8, C.I. 42000, Basic Green 4, 500g, 0.9 to 1.15, 13, 745, 219-441-7, 927.03, Pure, Danger | CAS: 2437-29-8 | C52H54N4O12 | 927.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2437-29-8
Molecular Formula C52H54N4O12
Molecular Weight (g/mol) 927.02
MDL Number MFCD00011766,MFCD00151209
InChI Key CNYGFPPAGUCRIC-UHFFFAOYSA-L
Synonym Basic Green 4, C.I. 42000
PubChem CID 2724411
IUPAC Name [4-[[4-(dimethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;2-hydroxy-2-oxoacetate;oxalic acid
SMILES OC(=O)C(O)=O.OC(=O)C([O-])=O.OC(=O)C([O-])=O.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C

Specifications

Absorbance 1650 to 1950 (at lambda max.)
Melting Point 164.0°C
Absorption Ratio 0.9 to 1.15
Infrared Spectrum Authentic
Lambda Max 620 to 625nm (in 50:50 IMS:water)
Loss on Drying 10% max. (105°C)
Packaging Glass bottle
Beilstein 13, 745
Merck Index 15, 5763
Color Green
Solubility Information Solubility in water: 60g/L (20°C). Other solubilities: soluble in alcohol, methanol, amyl alcohol
Quantity 500 g
Physical Form Crystalline Powder or Crystals With Metal Luster
Formula Weight 927.03
Grade Pure
Chemical Name or Material Malachite Green oxalate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Suspected of damaging the unborn child.
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 219-441-7
RTECSNumber BQ1190000

RUO – Research Use Only