missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Malachite Green oxalate, pure, certified

Catalog No. AC413491000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC413491000 100 g Glass bottle
AC413490250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC413491000 Supplier Thermo Scientific Chemicals Supplier No. 413491000
Only null left

CAS: 2437-29-8 | C52H54N4O12 | 927.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2437-29-8
Molecular Formula C52H54N4O12
Molecular Weight (g/mol) 927.02
MDL Number MFCD00011766,MFCD00151209
InChI Key CNYGFPPAGUCRIC-UHFFFAOYSA-L
Synonym Basic Green 4, C.I. 42000
PubChem CID 2724411
IUPAC Name [4-[[4-(dimethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;2-hydroxy-2-oxoacetate;oxalic acid
SMILES OC(=O)C(O)=O.OC(=O)C([O-])=O.OC(=O)C([O-])=O.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C

Specifications

Absorption Ratio 1.00 to 1.15
Dye Content 90% min.
Beilstein 13, 745
Merck Index 15, 5763
Infrared Spectrum Authentic
Lambda Max 614 to 620nm (in water)
Formula Weight 927.03
Grade Pure
Packaging Glass bottle
Vapor Pressure Vapor Pressure: Negligible
Visual Transition Interval from pH 0.0 (yellow) to pH 2.0 (green) from pH 11.6 (blue-green) to pH 14.0 (colorless)
Color Green
Quantity 100 g
Physical Form Metallic To Crystalline Powder or Crystals
Chemical Name or Material Malachite Green oxalate, Pure
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Toxic if swallowed. Suspected of damaging the unborn child. Causes serious eye damage. Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Use personal protective equipment as required. Avoid release to the environment.

RUO – Research Use Only