accessibility menu, dialog, popup

UserName

CAS RN 23582-03-8

IUPAC Name: (2-{bis[2-(diphenylphosphanyl)ethyl]phosphanyl}ethyl)diphenylphosphane
Synonyms: (2-{bis[2-(diphenylphosphanyl)ethyl]phosphanyl}ethyl)diphenylphosphane
Molecular Weight (g/mol): 670.69
Molecular Formula: C42H42P4
InChi Key: TVLNGWSWPKIYAO-UHFFFAOYSA-N
SMILES: C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CCP(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)