missing translation for 'onlineSavingsMsg'
Learn More

Tris[2-(diphenylphosphino)ethyl]phosphine, 97%

Catalog No. AC316830250
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC316830250 25 g
AC316830010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC316830250 Supplier Thermo Scientific Chemicals Supplier No. 316830250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 23582-03-8 | C42H42P4 | 670.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23582-03-8
Molecular Formula C42H42P4
Molecular Weight (g/mol) 670.69
MDL Number MFCD00010009
InChI Key TVLNGWSWPKIYAO-UHFFFAOYSA-N
Synonym tris 2-diphenylphosphino ethyl phosphine, tris 2-diphenylphosphinoethyl phosphine, tetraphos-li, 2-bis 2-diphenylphosphanyl ethyl phosphanyl ethyl diphenylphosphane, acmc-20ajaw, pubchem6541, chembl65725, tris 2-diphenylphosphanylethyl phosphane
PubChem CID 98777
IUPAC Name tris(2-diphenylphosphanylethyl)phosphane
SMILES C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CCP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 134.0°C to 139.0°C
Assay Percent Range 97%
Linear Formula [(C6H5)2PCH2CH2]3P
Quantity 25 g
Formula Weight 670.69
Percent Purity 97%
Chemical Name or Material Tris[2-(diphenylphosphino)ethyl]phosphine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 245-754-3

RUO – Research Use Only