missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris[2-(diphenylphosphino)ethyl]phosphine, 97%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| CAS | 23582-03-8 |
| Assay Percent Range | 97% |
| Packaging | Glass bottle |
| Molecular Formula | C42H42P4 |
| Linear Formula | [(C6H5)2PCH2CH2]3P |
| Quantity | 5g |
| MDL Number | MFCD00010009 |
| Synonym | tris 2-diphenylphosphino ethyl phosphine, tris 2-diphenylphosphinoethyl phosphine, tetraphos-li, 2-bis 2-diphenylphosphanyl ethyl phosphanyl ethyl diphenylphosphane, acmc-20ajaw, pubchem6541, chembl65725, tris 2-diphenylphosphanylethyl phosphane |
| InChI Key | TVLNGWSWPKIYAO-UHFFFAOYSA-N |
| SMILES | C1=CC=C(C=C1)P(CCP(CCP(C2=CC=CC=C2)C3=CC=CC=C3)CCP(C4=CC=CC=C4)C5=CC=CC=C5)C6=CC=CC=C6 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement: Harmful in contact with skin. Harmful if inhaled. Harmful if swallowed. Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation. |
| Precautionary Statement | GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 245-754-3 |
RUO – Research Use Only