accessibility menu, dialog, popup

UserName

HOOFCl

CAS RN 2252-51-9

IUPAC Name: 2-chloro-4-fluorobenzoic acid
Synonyms: 2-chloro-4-fluorobenzoic acid
Molecular Weight (g/mol): 174.56
Molecular Formula: C7H4ClFO2
InChi Key: GRPWQLDSGNZEQE-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(F)C=C1Cl
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)