missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorobenzoic Acid 98.0+%, TCI America™

Catalog No. C13505G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13505G 5 g
C135025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13505G Supplier TCI America Supplier No. C13505G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2252-51-9
Molecular Formula C7H4ClFO2
Molecular Weight (g/mol) 174.555
MDL Number MFCD00010615
InChI Key GRPWQLDSGNZEQE-UHFFFAOYSA-N
Synonym benzoic acid, 2-chloro-4-fluoro, 2-chloro-4-fluoro-benzoic acid, 2-chloro-4-fluor obenzoic acid, pubchem1370, acmc-209fx5, ksc207e4n, rarechem al bo 0405, 2-chloro 4-fluoro benzoic acid, 2-chloro-4-fluoro benzoic acid, 4-fluoro-2-chloro-benzoic acid
PubChem CID 75259
IUPAC Name 2-chloro-4-fluorobenzoic acid
SMILES C1=CC(=C(C=C1F)Cl)C(=O)O

Specifications

Melting Point 185°C
Color White
Quantity 5 g
Formula Weight 174.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-fluorobenzoic Acid