accessibility menu, dialog, popup

UserName

CAS RN 22504-50-3

HSOOOOSH

CAS RN 22504-50-3

IUPAC Name: 2-[(3-sulfanylpropanoyl)oxy]ethyl 3-sulfanylpropanoate
Synonyms: 2-[(3-sulfanylpropanoyl)oxy]ethyl 3-sulfanylpropanoate
Molecular Weight (g/mol): 238.32
Molecular Formula: C8H14O4S2
InChi Key: HAQZWTGSNCDKTK-UHFFFAOYSA-N
SMILES: SCCC(=O)OCCOC(=O)CCS
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)