missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol Bis(3-mercaptopropionate) (Purified) 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 g |
| Formula Weight | 238.32 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Liquid |
| Chemical Name or Material | Ethylene Glycol Bis(3-mercaptopropionate) (Purified) |
Safety and Handling
| TSCA | Yes |