missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Bis(3-mercaptopropionate) 97.0+%, TCI America™

Catalog No. E1269500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E1269500G 500 g
E126925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E1269500G Supplier TCI America Supplier No. E1269500G
Only null left

Chemical Identifiers

CAS 22504-50-3
Molecular Formula C8H14O4S2
Molecular Weight (g/mol) 238.316
MDL Number MFCD00053520
InChI Key HAQZWTGSNCDKTK-UHFFFAOYSA-N
Synonym Ethylene Bis(3-mercaptopropionate), Glycol Bis(3-mercaptopropionate)
PubChem CID 89735
IUPAC Name 2-(3-sulfanylpropanoyloxy)ethyl 3-sulfanylpropanoate
SMILES C(CS)C(=O)OCCOC(=O)CCS

Specifications

Color Yellow
Quantity 500 g
Formula Weight 238.32
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Ethylene Glycol Bis(3-mercaptopropionate)