accessibility menu, dialog, popup

UserName

CH3H3CSiClCH3H3C

CAS RN 2227-29-4

IUPAC Name: chlorobis(propan-2-yl)silyl
Synonyms: chlorodiisopropylsilyl
Molecular Weight (g/mol): 149.71
Molecular Formula: C6H14ClSi
InChi Key: IGSUJBNDAWQLST-UHFFFAOYSA-N
SMILES: CC(C)[Si](Cl)C(C)C
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)