missing translation for 'onlineSavingsMsg'
Learn More

Chlorodiisopropylsilane 95.0+%, TCI America™

Catalog No. C14925ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14925ML 5 mL
C149225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14925ML Supplier TCI America Supplier No. C14925ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2227-29-4
Molecular Formula C6H14ClSi
Molecular Weight (g/mol) 149.71
MDL Number MFCD00054896
InChI Key IGSUJBNDAWQLST-UHFFFAOYSA-N
Synonym chlorodiisopropylsilane, diisopropylchlorosilane, unii-dvg177547j, silane, chloro diisopropyl, zlchem 966, chlorodiisopropylsilyl, chlorobis propan-2-yl silane, chloro-di propan-2-yl silicon
PubChem CID 6365034
IUPAC Name chlorobis(propan-2-yl)silyl
SMILES CC(C)[Si](Cl)C(C)C

Specifications

Color Colorless
Boiling Point 143°C
Quantity 5 mL
UN Number 2985
Formula Weight 150.72
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Chlorodiisopropylsilane
EINECSNumber (2)-2041
TSCA Yes