accessibility menu, dialog, popup

UserName

CAS RN 21800-83-9

CH3(S)(R)O(R)(S)O

CAS RN 21800-83-9

IUPAC Name: (3aS,3bR,9aR,9bS)-11a-ethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-1,7-dione
Synonyms: (3aS,3bR,9aR,9bS)-11a-ethyl-2H,3H,3aH,3bH,4H,5H,8H,9H,9aH,9bH,10H,11H-cyclopenta[a]phenanthrene-1,7-dione
Molecular Weight (g/mol): 286.42
Molecular Formula: C19H26O2
InChi Key: SBLHOJQRZNGHLQ-YLVOMZDISA-N
SMILES: CCC12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CCC2=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results