accessibility menu, dialog, popup

UserName

H3CONONH

CAS RN 21642-98-8

IUPAC Name: 4-methoxy-2-oxo-1,2-dihydropyridine-3-carbonitrile
Synonyms: 4-methoxy-2-oxo-1H-pyridine-3-carbonitrile
Molecular Weight (g/mol): 150.14
Molecular Formula: C7H6N2O2
InChi Key: MWGIDWPSRDMIQN-UHFFFAOYSA-N
SMILES: COC1=C(C#N)C(=O)NC=C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)