missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Cyano-4-methoxy-2-pyridone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 278°C |
| Color | White-Yellow |
| Quantity | 5 g |
| UN Number | 3439 |
| Formula Weight | 150.14 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Cyano-4-methoxy-2-pyridone |
Safety and Handling
| TSCA | No |