accessibility menu, dialog, popup

UserName

CAS RN 2142-73-6

IUPAC Name: 1-(2,5-dimethylphenyl)ethan-1-one
Synonyms: 1-(2,5-dimethylphenyl)ethanone
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: AWKBVLVKQQRRFQ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(C)=CC=C1C
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)