accessibility menu, dialog, popup

UserName

CAS RN 21324-40-3

IUPAC Name: lithium(1+) hexafluoro-λ⁵-phosphanuide
Synonyms: lithium hexafluorophosphate
Molecular Weight (g/mol): 151.9
Molecular Formula: F6LiP
InChi Key: AXPLOJNSKRXQPA-UHFFFAOYSA-N
SMILES: [Li+].F[P-](F)(F)(F)(F)F
Molecular Weight (g/mol) 
  • (5)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (6)
  • (5)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)