missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Hexafluorophosphate de lithium
Description
Hexafluorophosphate de lithium
Spécifications
Spécifications
| Melting Point | 200°C |
| CAS | 21324-40-3 |
| Quantity | 25 g |
| Molecular Formula | F6LiP |
| Linear Formula | LiPF6 |
| MDL Number | MFCD00011096 |
| Synonym | Lithium phosphorus fluoride |
| Molecular Weight (g/mol) | 151.91 |
| Percent Purity | ≥99.99% trace metals basis |