missing translation for 'onlineSavingsMsg'
Learn More

Lithium hexafluorophosphate, 98%

Catalog No. AA1152918
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1152918 50 g
AA1152903 1 g
AA1152909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1152918 Supplier Thermo Scientific Chemicals Supplier No. 01152918
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used As An Electrolyte in Li-Ion Batteries | CAS: 21324-40-3 | F6LiP | 151.904 g/mol

Lithium sulfate is used to treat bipolar disorder. It is also used in ultrasound-type non-destructive testing because they are very efficient sound generators. It is also used in building blocks and chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 21324-40-3
Molecular Formula F6LiP
Molecular Weight (g/mol) 151.904
MDL Number MFCD00011096
InChI Key AXPLOJNSKRXQPA-UHFFFAOYSA-N
Synonym lithium hexafluorophosphate, lithium hexafluorophosphate v, phosphate 1-, hexafluoro-, lithium, phosphate 1-, hexafluoro-, lithium 1:1, lithiumhexafluorophosphate, lithium hexafluorophosphate 1-, lipf6, lithium hexafluoroposphate, acmc-209fj7, ksc492g4h
PubChem CID 23688915
IUPAC Name lithium;hexafluorophosphate
SMILES [Li+].F[P-](F)(F)(F)(F)F

Specifications

Melting Point 200°C (decomposition)
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 98%
UN Number 2923
Solubility Information Soluble in water.
Formula Weight 151.9
Odor Odorless
Sensitivity Moisture sensitive
Appearance White powder
Density 1.5 g/cm3 at 20°C
Chemical Name or Material Lithium Hexafluorophosphate
Show More Show Less
Hazard Category H301-H314-H335-H372
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation DOT Class: 8; Packaging Group: I
EINECSNumber 244-334-7
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only