accessibility menu, dialog, popup

UserName

CAS RN 20559-55-1

IUPAC Name: methyl N-(6-propoxy-1H-1,3-benzodiazol-2-yl)carbamate
Synonyms: oxibendazole
Molecular Weight (g/mol): 249.27
Molecular Formula: C12H15N3O3
InChi Key: RAOCRURYZCVHMG-UHFFFAOYSA-N
SMILES: CCCOC1=CC=C2N=C(NC(=O)OC)NC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results