missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Oxibendazole, MilliporeSigma™ Supelco™

Catalog No. 111013488
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013488 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013488 Supplier Merck Emd Millipore Supplier No. 32924100MG

Sigma-Aldrich offers oxibendazole for a variety of applications that include, but are not limited to, environmental, food and beverage, forensics, petrochemical and pharmaceutical analyses.

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 20559-55-1
Physical Form Neat
Linear Formula C12H15N3O3
MDL Number MFCD00133728
Synonym Methyl (5-propoxy-1 H-benzimidazol-2-yl)carbamate
Type Analytical Standard
Quantity 100 mg
Formula Weight 249.27
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH