accessibility menu, dialog, popup

UserName

CAS RN 19771-63-2

IUPAC Name: (4R)-2-oxo-1,3-thiazolidine-4-carboxylic acid
Synonyms: oxothiazolidinecarboxylic acid
Molecular Weight (g/mol): 147.15
Molecular Formula: C4H5NO3S
InChi Key: BMLMGCPTLHPWPY-REOHCLBHSA-N
SMILES: OC(=O)[C@@H]1CSC(=O)N1
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results