missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Oxothiazolidine-4-carboxylic acid, Thermo Scientific Chemicals

Catalog No. AAJ6225403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6225403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6225403 Supplier Thermo Scientific Chemicals Supplier No. J6225403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reagent for hepatic glutathione studies

Reagent for hepatic glutathione studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19771-63-2
Molecular Formula C4H5NO3S
Molecular Weight (g/mol) 147.148
MDL Number MFCD00066092
InChI Key BMLMGCPTLHPWPY-REOHCLBHSA-N
Synonym procysteine, r-2-oxothiazolidine-4-carboxylic acid, 4r-2-oxo-1,3-thiazolidine-4-carboxylic acid, l-2-oxothiazolidine-4-carboxylate, oxothiazolidinecarboxylic acid, l-2-oxothiazolidine-4-carboxylic acid, 4-thiazolidinecarboxylic acid, 2-oxo-, 4r, ccris 7672, oxothiazolidine carboxylate, l, r---2-oxothiazolidine-4-carboxylic acid
PubChem CID 72390
IUPAC Name (4R)-2-oxo-1,3-thiazolidine-4-carboxylic acid
SMILES C1C(NC(=O)S1)C(=O)O

Specifications

Melting Point 174°C (decomposition)
Color White
Beilstein 4179169
Merck Index 14,6950
Solubility Information Soluble in water
Quantity 1 g
Formula Weight 147.15
Physical Form Powder
Chemical Name or Material (R)-(-)-2-Oxothiazolidine-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber XJ5426650
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only