missing translation for 'onlineSavingsMsg'
Learn More

L-2-Thiazolidinone-4-carboxylic Acid 97.0+%, TCI America™

Catalog No. T12235G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12235G 5 g
T12231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T12235G Supplier TCI America Supplier No. T12235G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19771-63-2
Molecular Formula C4H5NO3S
Molecular Weight (g/mol) 147.148
MDL Number MFCD00066092
InChI Key BMLMGCPTLHPWPY-REOHCLBHSA-N
Synonym procysteine, r-2-oxothiazolidine-4-carboxylic acid, 4r-2-oxo-1,3-thiazolidine-4-carboxylic acid, l-2-oxothiazolidine-4-carboxylate, oxothiazolidinecarboxylic acid, l-2-oxothiazolidine-4-carboxylic acid, 4-thiazolidinecarboxylic acid, 2-oxo-, 4r, ccris 7672, oxothiazolidine carboxylate, l, r---2-oxothiazolidine-4-carboxylic acid
PubChem CID 72390
IUPAC Name (4R)-2-oxo-1,3-thiazolidine-4-carboxylic acid
SMILES C1C(NC(=O)S1)C(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 147.15
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-2-Thiazolidinone-4-carboxylic Acid
RTECSNumber XJ5426650
TSCA No