accessibility menu, dialog, popup

UserName

CH3BOH3CH3CH3CCH3O

CAS RN 195062-57-8

IUPAC Name: 4,4,5,5-tetramethyl-2-(4-methylphenyl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(4-methylphenyl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 218.1
Molecular Formula: C13H19BO2
InChi Key: GKSSEDDAXXEPCP-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results