missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4,5,5-Tetramethyl-2-(p-tolyl)-1,3,2-dioxaborolane 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 54°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 218.10 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4,4,5,5-Tetramethyl-2-(p-tolyl)-1,3,2-dioxaborolane |
Safety and Handling
| TSCA | No |