accessibility menu, dialog, popup

UserName

HOONHOO

CAS RN 1947-00-8

IUPAC Name: 6-{[(benzyloxy)carbonyl]amino}hexanoic acid
Synonyms: 6-{[(benzyloxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol): 265.31
Molecular Formula: C14H19NO4
InChi Key: RXQDBVWDABAAHL-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCNC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)