missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-6-aminohexanoic Acid 98.0+%, TCI America™

Catalog No. C27575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27575G 5 g
C275725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27575G Supplier TCI America Supplier No. C27575G

Chemical Identifiers

CAS 1947-00-8
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.309
MDL Number MFCD00004423
InChI Key RXQDBVWDABAAHL-UHFFFAOYSA-N
Synonym z-6-aminohexanoic acid, z-epsilon-acp-oh, 6-benzyloxy carbonyl amino hexanoic acid, n-benzyloxycarbonyl-6-aminocaproic acid, cbz-, a-aminocaproic acid, n-cbz-6-aminohexanoic acid, n-benzyloxycarbonyl-6-aminohexanoic acid, n-carbobenzoxy-epsilon-aminocaproic acid, 6-phenylmethoxy carbonyl amino hexanoic acid
PubChem CID 74758
IUPAC Name 6-(phenylmethoxycarbonylamino)hexanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NCCCCCC(=O)O

Specifications

Melting Point 58°C
Color White
Quantity 5 g
Formula Weight 265.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-6-aminohexanoic Acid
TSCA No
Recommended Storage Refrigerator