missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-6-aminohexanoic acid, 98%

Catalog No. AAL0899714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0899714 25 g
AAL0899706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0899714 Supplier Thermo Scientific Chemicals Supplier No. L0899714

CAS: 1947-00-8 | C14H19NO4 | 265.309 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1947-00-8
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.309
MDL Number MFCD00004423
InChI Key RXQDBVWDABAAHL-UHFFFAOYSA-N
Synonym z-6-aminohexanoic acid, z-epsilon-acp-oh, 6-benzyloxy carbonyl amino hexanoic acid, n-benzyloxycarbonyl-6-aminocaproic acid, cbz-, a-aminocaproic acid, n-cbz-6-aminohexanoic acid, n-benzyloxycarbonyl-6-aminohexanoic acid, n-carbobenzoxy-epsilon-aminocaproic acid, 6-phenylmethoxy carbonyl amino hexanoic acid
PubChem CID 74758
IUPAC Name 6-(phenylmethoxycarbonylamino)hexanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NCCCCCC(=O)O

Specifications

Melting Point 53°C to 57°C
Quantity 25 g
Beilstein 2288508
Formula Weight 265.31
Percent Purity 99%
Chemical Name or Material N-Benzyloxycarbonyl-6-aminohexanoic acid
EINECSNumber 217-748-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only