accessibility menu, dialog, popup

UserName

CAS RN 18908-66-2

IUPAC Name: 3-(bromomethyl)heptane
Synonyms: 3-(bromomethyl)heptane
Molecular Weight (g/mol): 193.13
Molecular Formula: C8H17Br
InChi Key: NZWIYPLSXWYKLH-UHFFFAOYNA-N
SMILES: CCCCC(CC)CBr
Boiling Point 
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (6)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (5)