missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl bromide, 96%, stab. with copper

Catalog No. AAB2353236
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2353236 500 g
AAB2353214 25 g
AAB2353222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2353236 Supplier Thermo Scientific Chemicals Supplier No. B2353236
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18908-66-2 | C8H17Br | 193.128 g/mol

2-Ethylhexyl bromide is used in the synthesis of methoxy-4-(2-ethylhexyloxy)benzene, 1-bromo-4-(2?-ethylhexyloxy)benzene and 3,6-diaryl substituted 9-alkylcarbazoles as hole transporting materials for various organic light emitting devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylhexyl bromide is used in the synthesis of methoxy-4-(2-ethylhexyloxy)benzene, 1-bromo-4-(2′-ethylhexyloxy)benzene and 3,6-diaryl substituted 9-alkylcarbazoles as hole transporting materials for various organic light emitting devices.

Solubility
Soluble in chloroform, ethyl acetate. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from heat, oxidizing agents.

Chemical Identifiers

CAS 18908-66-2
Molecular Formula C8H17Br
Molecular Weight (g/mol) 193.128
MDL Number MFCD00000220
InChI Key NZWIYPLSXWYKLH-UHFFFAOYSA-N
Synonym 2-ethylhexyl bromide, 3-bromomethyl heptane, heptane, 3-bromomethyl, 1-bromo-2-ethylhexane, 1-bromo iso octane, 2-ethyl-bromohexane, 3-bromomethyl-heptane, 1-bromo-2 ethylhexane, 2-ethylhexylhydrobromide
PubChem CID 86804
IUPAC Name 3-(bromomethyl)heptane
SMILES CCCCC(CC)CBr

Specifications

Density 1.086
Boiling Point 94°C to 95°C (35 mmHg)
Flash Point 69°C (156°F)
Refractive Index 1.4538
Quantity 500 g
Beilstein 1098313
Solubility Information Soluble in chloroform,ethyl acetate. Insoluble in water.
Formula Weight 193.13
Percent Purity 96%
Chemical Name or Material 2-Ethylhexyl bromide, stabilized with copper
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 242-659-9
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only