missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-ethylhexane 97.0+%, TCI America™

Catalog No. B0596500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0596500ML 500 mL
B059625ML 25 mL
B0596100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0596500ML Supplier TCI America Supplier No. B0596500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18908-66-2
Molecular Formula C8H17Br
Molecular Weight (g/mol) 193.128
MDL Number MFCD00000220
InChI Key NZWIYPLSXWYKLH-UHFFFAOYSA-N
Synonym 2-ethylhexyl bromide, 3-bromomethyl heptane, heptane, 3-bromomethyl, 1-bromo-2-ethylhexane, 1-bromo iso octane, 2-ethyl-bromohexane, 3-bromomethyl-heptane, 1-bromo-2 ethylhexane, 2-ethylhexylhydrobromide
PubChem CID 86804
IUPAC Name 3-(bromomethyl)heptane
SMILES CCCCC(CC)CBr

Specifications

Color Yellow
Boiling Point 156°C
Quantity 500 mL
Formula Weight 193.13
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-ethylhexane
EINECSNumber (2)-0067
TSCA Yes