accessibility menu, dialog, popup

UserName

CAS RN 18416-07-4

IUPAC Name: dichlorodioctylsilane
Synonyms: dichlorodioctylsilane
Molecular Weight (g/mol): 325.43
Molecular Formula: C16H34Cl2Si
InChi Key: CVAGYCLEIYGJQT-UHFFFAOYSA-N
SMILES: CCCCCCCC[Si](Cl)(Cl)CCCCCCCC
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results