missing translation for 'onlineSavingsMsg'
Learn More

Dichlorodi-n-octylsilane 96.0+%, TCI America™

Catalog No. D43361G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D43361G Supplier TCI America Supplier No. D43361G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18416-07-4
Molecular Formula C16H34Cl2Si
Molecular Weight (g/mol) 325.433
MDL Number MFCD01074472
InChI Key CVAGYCLEIYGJQT-UHFFFAOYSA-N
Synonym di-n-octyldichlorosilane, dichloro dioctyl silane, silane, dichlorodioctyl, dioctyldichlorosilane, dioctyldichlorsilan, dichlorodi-n-octylsilane, di-n-octyldichlorosilane 10g
PubChem CID 248429
IUPAC Name dichloro(dioctyl)silane
SMILES CCCCCCCC[Si](CCCCCCCC)(Cl)Cl

Specifications

Color Colorless
Boiling Point 150°C
Quantity 1 g
UN Number 2987
Formula Weight 325.43
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Dichlorodi-n-octylsilane
EINECSNumber (2)-2041
TSCA No