missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-n-octyldichlorosilane, 97%
Description
Di-n-octyldichlorosilane is used in the synthesis of organic photovoltaics. It is also used in medicine field, electronic industry and polymer material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 145°C (0.2mmHg) |
| Color | Colorless to Yellow |
| Physical Form | Liquid |
| Quantity | 50 g |
| Sensitivity | Moisture sensitive |
| Density | 0.940 g/mL |
| Linear Formula | [CH3(CH2)7]2SiCl2 |
| UN Number | UN2987 |
| Solubility Information | Sparingly Soluble (8.8E-6g/L) at 25°C. |
| Formula Weight | 325.44 |
| Show More |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only