accessibility menu, dialog, popup

UserName

CAS RN 17912-87-7

CH3(S)O(S)OOOHOHOHOHOOH(R)HO(R)HO(R)OH

CAS RN 17912-87-7

IUPAC Name: 5,7-dihydroxy-3-{[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one
Synonyms: myricitrin myricetin 3-rhamnoside rhamnoside,myricetin-3 myricitrin (8CI) myricitrine myricetol 3-rhamnoside myricetin 3-O-α-L-rhamnoside myricitroside myricetin 3-O-rhamnoside
Molecular Weight (g/mol): 464.38
Molecular Formula: C21H20O12
InChi Key: DCYOADKBABEMIQ-OWMUPTOHSA-N
SMILES: C[C@@H]1O[C@@H](OC2=C(OC3=CC(O)=CC(O)=C3C2=O)C2=CC(O)=C(O)C(O)=C2)[C@H](O)[C@H](O)[C@H]1O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results