missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Myricitrin, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002286
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002286 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002286 Supplier Merck Emd Millipore Supplier No. PHL8020920MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 17912-87-7
Product Line phyproof
Linear Formula C21H20O12
MDL Number MFCD00016930
Synonym 3,3′,4′,5,5′,7-Hexahydroxyflavone 3-O-rhamnoside; Myricetin 3-O-alpha-L-rhamnopyranoside; Myricetin 3-O-rhamnoside; Myricitroside
Type Reference Substance
Quantity 20 mg
Formula Weight 464.38
Percent Purity ≥95% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG