missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myricitrine, ≥95% (HPLC), MilliporeSigma™ Supelco™
Spécifications
Spécifications
| CAS | 17912-87-7 |
| Physical Form | Sympa |
| Linear Formula | C21H20O12 |
| MDL Number | MFCD00016930 |
| Synonym | 3,3′,4′,5,5′,7-Hexahydroxyflavone 3-O-rhamnoside; Myricetin 3-O-alpha-L-rhamnopyranoside; Myricetin 3-O-rhamnoside; Myricitroside |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 464.38 |
| Percent Purity | ≥95% (HPLC) |
| Grade | Norme analytique |