accessibility menu, dialog, popup

UserName

CH3H2COOOOCH3

CAS RN 177080-66-9

IUPAC Name: 4-methyl-2-oxooxan-4-yl 2-methylprop-2-enoate
Synonyms: 4-methyl-2-oxooxan-4-yl 2-methylprop-2-enoate
Molecular Weight (g/mol): 198.22
Molecular Formula: C10H14O4
InChi Key: JANRUIBBIKVUHU-UHFFFAOYNA-N
SMILES: CC(=C)C(=O)OC1(C)CCOC(=O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results