missing translation for 'onlineSavingsMsg'
Learn More

Mevalonic Lactone Methacrylate (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. M22611G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M22611G Supplier TCI America Supplier No. M22611G
Only null left

Chemical Identifiers

CAS 177080-66-9
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.218
InChI Key JANRUIBBIKVUHU-UHFFFAOYSA-N
Synonym Methacrylic Acid 4-Methyl-2-oxotetrahydro-2H-pyran-4-yl Ester, 4-Methyl-2-oxotetrahydro-2H-pyran-4-yl Methacrylate
PubChem CID 9964364
IUPAC Name (4-methyl-2-oxooxan-4-yl) 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC1(CCOC(=O)C1)C

Specifications

Quantity 1 g
Formula Weight 198.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Mevalonic Lactone Methacrylate (stabilized with MEHQ)
TSCA No
Recommended Storage Freezer